Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
haf-1
Entrez Gene ID
178522
Gene Name
HAlF transporter (PGP related)
Gene Aliases
CELE_C30H6.6
Chromosome Location
Chr.IV: 17376427 - 17379322 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070697.6 NP_503098.2 8 - 9 1845 129

View Full Product Details