Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cul-3
Entrez Gene ID
178547
Gene Name
protein-coding
Gene Aliases
CELE_Y108G3AL.1
Chromosome Location
Chr.V: 177417 - 185172 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313285.3 NP_001300214.1 2 - 3 1428 71
NM_070750.7 NP_503151.1 4 - 5 1720 71

View Full Product Details