Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dyci-1
Entrez Gene ID
177440
Gene Name
protein-coding
Gene Aliases
CELE_C17H12.1
Chromosome Location
Chr.IV: 6779774 - 6785272 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372852.3 NP_001360494.1 7 - 7 1204 105
NM_001383110.1 6 - 7 1117 105

View Full Product Details