Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dyci-1
Entrez Gene ID
177440
Gene Name
protein-coding
Gene Aliases
CELE_C17H12.1
Chromosome Location
Chr.IV: 6779774 - 6785272 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372852.3 NP_001360494.1 10 - 11 1757 76
NM_001383110.1 9 - 10 1670 76

View Full Product Details