Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-50
Entrez Gene ID
177478
Gene Name
protein-coding
Gene Aliases
CELE_Y43B11AL.2
Chromosome Location
Chr.IV: 7057977 - 7059911 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380302.1 NP_001367449.1 3 - 4 357 115
NM_001380303.1 NP_001367448.1 3 - 4 338 115

View Full Product Details