Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-15
Entrez Gene ID
178558
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_F33E11.1
Chromosome Location
Chr.V: 312721 - 323650 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070773.2 NP_503174.1 6 - 7 1013 116
NR_132474.1 5 - 6 1321 116

View Full Product Details