Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fmo-5
Entrez Gene ID
178596
Gene Name
protein-coding
Gene Aliases
CELE_H24K24.5
Chromosome Location
Chr.V: 899304 - 903665 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070951.5 NP_503352.1 3 - 4 518 157
GenBank mRNA AJ582073.1 - 3 - 4 432 157

View Full Product Details