Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fmo-5
Entrez Gene ID
178596
Gene Name
protein-coding
Gene Aliases
CELE_H24K24.5
Chromosome Location
Chr.V: 899304 - 903665 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313203.3 NP_001300132.1 1 - 2 214 108
NM_070951.5 NP_503352.1 6 - 7 1346 108
GenBank mRNA AJ582073.1 - 6 - 7 1260 108

View Full Product Details