Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srg-57
Entrez Gene ID
182842
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_C24B9.14
Chromosome Location
Chr.V: 2732177 - 2734460 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071342.5 NP_503743.3 1 - 2 194 162

View Full Product Details