Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-57
Entrez Gene ID
182842
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_C24B9.14
Chromosome Location
Chr.V: 2732177 - 2734460 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071342.5 NP_503743.3 2 - 3 410 154

View Full Product Details