Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-24
Entrez Gene ID
3565787
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C24B9.16
Chromosome Location
Chr.V: 2748152 - 2750106 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028499.2 NP_001023670.2 2 - 3 205 96

View Full Product Details