Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-57
Entrez Gene ID
191954
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_T03D3.14
Chromosome Location
Chr.V: 2854393 - 2855573 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071371.1 NP_503772.1 4 - 5 825 88

View Full Product Details