Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-24
Entrez Gene ID
185415
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_F37B4.9
Chromosome Location
Chr.V: 2884711 - 2886425 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071383.1 NP_503784.1 1 - 2 458 96

View Full Product Details