Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-33C9
Entrez Gene ID
178752
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_C50H11.15
Chromosome Location
Chr.V: 3091812 - 3094561 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071445.5 NP_503846.1 2 - 3 641 83

View Full Product Details