Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-68
Entrez Gene ID
191985
Gene Name
protein-coding
Gene Aliases
CELE_Y73C8C.6
Chromosome Location
Chr.V: 3106228 - 3107550 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071450.1 NP_503851.1 1 - 2 489 105

View Full Product Details