Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srab-21
Entrez Gene ID
188653
Gene Name
Serpentine Receptor, class AB (class A-like)
Gene Aliases
CELE_T20D4.18
Chromosome Location
Chr.V: 3422949 - 3424441 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071558.2 NP_503959.2 3 - 4 645 151

View Full Product Details