Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-9
Entrez Gene ID
191934
Gene Name
protein-coding
Gene Aliases
CELE_T20C4.1
Chromosome Location
Chr.V: 3431940 - 3433751 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322515.1 NP_001309651.1 2 - 3 357 108
NM_001322516.1 NP_001309653.1 1 - 2 81 108
NM_001322517.1 NP_001309652.1 1 - 2 81 108
NM_071562.2 NP_503963.1 2 - 3 676 108

View Full Product Details