Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-12
Entrez Gene ID
186714
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_H10D18.2
Chromosome Location
Chr.V: 3812591 - 3813437 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071655.3 NP_504056.1 3 - 4 537 64

View Full Product Details