Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-77
Entrez Gene ID
186920
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_K03B4.5
Chromosome Location
Chr.V: 4666745 - 4668736 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071856.3 NP_504257.3 4 - 5 740 165

View Full Product Details