Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hil-4
Entrez Gene ID
178866
Gene Name
Histone H1.4
Gene Aliases
CELE_C18G1.5
Chromosome Location
Chr.V: 4754313 - 4755453 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383299.1 1 - 2 317 99
GenBank mRNA AF005371.1 - 1 - 2 297 99

View Full Product Details