Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-48
Entrez Gene ID
178940
Gene Name
Putative UDP-glucuronosyltransferase ugt-48
Gene Aliases
CELE_C18C4.3
Chromosome Location
Chr.V: 5545650 - 5548322 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072063.7 NP_504464.2 8 - 9 1431 92

View Full Product Details