Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-15
Entrez Gene ID
191804
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C04E6.10
Chromosome Location
Chr.V: 5904917 - 5906352 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072137.4 NP_504538.1 1 - 2 207 115

View Full Product Details