Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-20
Entrez Gene ID
191809
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_Y38A10A.1
Chromosome Location
Chr.V: 6055589 - 6056743 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072168.1 NP_504569.1 1 - 2 188 129

View Full Product Details