The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083280.3 | NP_001076749.1 | 10 - 11 | 1418 | 123 |
NM_001373040.1 | NP_001360140.1 | 10 - 11 | 1418 | 123 | |
NM_001383319.1 | 10 - 11 | 1396 | 123 | ||
GenBank mRNA | AF338767.1 | - | 10 - 11 | 1405 | 123 |