Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-83
Entrez Gene ID
179033
Gene Name
protein-coding
Gene Aliases
CELE_W01A11.3
Chromosome Location
Chr.V: 6465063 - 6487883 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083280.3 NP_001076749.1 13 - 14 2062 65
NM_001373040.1 NP_001360140.1 13 - 14 2062 65
NM_001383319.1 13 - 14 2040 65
GenBank mRNA AF338767.1 - 13 - 14 2049 65

View Full Product Details