Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dbl-1
Entrez Gene ID
179068
Gene Name
TGF_BETA_2 domain-containing protein
Gene Aliases
CELE_T25F10.2
Chromosome Location
Chr.V: 6758135 - 6760418 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072308.8 NP_504709.1 4 - 5 902 64
GenBank mRNA AF004395.1 - 4 - 5 860 64

View Full Product Details