Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-30
Entrez Gene ID
191851
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_ZC404.12
Chromosome Location
Chr.V: 6769634 - 6771745 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072313.6 NP_504714.1 4 - 5 895 76

View Full Product Details