Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-16
Entrez Gene ID
188256
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T07H8.1
Chromosome Location
Chr.V: 6976347 - 6977387 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072362.2 NP_504763.2 1 - 2 43 83

View Full Product Details