Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-83
Entrez Gene ID
191875
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_F09G2.7
Chromosome Location
Chr.V: 7164510 - 7165895 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072418.1 NP_504819.1 1 - 2 172 119

View Full Product Details