Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-7
Entrez Gene ID
184314
Gene Name
C-type LECtin
Gene Aliases
CELE_F10G2.3
Chromosome Location
Chr.V: 7328392 - 7330002 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072464.5 NP_504865.1 1 - 2 230 151

View Full Product Details