Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-24
Entrez Gene ID
182178
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C03G6.16
Chromosome Location
Chr.V: 7377897 - 7379484 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072486.1 NP_504887.1 6 - 7 811 107

View Full Product Details