Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gst-41
Entrez Gene ID
179127
Gene Name
Glutathione S-Transferase
Gene Aliases
CELE_R13D7.7
Chromosome Location
Chr.V: 7398396 - 7399302 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072493.6 NP_504894.1 2 - 3 243 78

View Full Product Details