Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-79
Entrez Gene ID
191873
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_C04F2.1
Chromosome Location
Chr.V: 7497740 - 7499128 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072529.2 NP_504930.1 3 - 4 876 138

View Full Product Details