Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-94
Entrez Gene ID
179149
Gene Name
protein-coding
Gene Aliases
CELE_C12D5.8
Chromosome Location
Chr.V: 7684052 - 7686386 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383341.1 4 - 5 429 70
GenBank mRNA AY204182.1 - 4 - 5 385 70

View Full Product Details