Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-29
Entrez Gene ID
185622
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_F41E6.14
Chromosome Location
Chr.V: 8631871 - 8635675 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072827.3 NP_505228.2 7 - 8 1174 184

View Full Product Details