Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glo-4
Entrez Gene ID
179306
Gene Name
X-linked retinitis pigmentosa GTPase regulator homolog
Gene Aliases
CELE_F07C3.4
Chromosome Location
Chr.V: 9234610 - 9243221 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072991.5 NP_505392.3 11 - 12 1897 69

View Full Product Details