Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-38
Entrez Gene ID
187651
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_R07B5.4
Chromosome Location
Chr.V: 9834463 - 9836088 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073137.2 NP_505538.1 1 - 2 471 79

View Full Product Details