Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-21
Entrez Gene ID
187652
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_R07B5.5
Chromosome Location
Chr.V: 9837979 - 9839508 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073138.5 NP_505539.2 6 - 7 764 96

View Full Product Details