Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gly-20
Entrez Gene ID
179562
Gene Name
GLYcosylation related
Gene Aliases
CELE_C03E10.4
Chromosome Location
Chr.V: 11282570 - 11286039 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073463.8 NP_505864.1 5 - 6 1400 152

View Full Product Details