Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhs-19
Entrez Gene ID
179578
Gene Name
DeHydrogenases, Short chain
Gene Aliases
CELE_T11F9.11
Chromosome Location
Chr.V: 11489048 - 11490482 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073514.7 NP_505915.1 3 - 4 547 113

View Full Product Details