Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-15
Entrez Gene ID
185260
Gene Name
GLoBin related
Gene Aliases
CELE_F35B12.8
Chromosome Location
Chr.V: 11616805 - 11618273 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073546.6 NP_505947.2 2 - 3 330 124

View Full Product Details