Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sel-11
Entrez Gene ID
179612
Gene Name
E3 ubiquitin-protein ligase hrd-1
Gene Aliases
CELE_F55A11.3
Chromosome Location
Chr.V: 11767328 - 11769902 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073568.6 NP_505969.1 5 - 6 1287 55

View Full Product Details