Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clh-6
Entrez Gene ID
179655
Gene Name
Chloride channel protein
Gene Aliases
CELE_R07B7.1
Chromosome Location
Chr.V: 12060233 - 12063368 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073621.5 NP_506022.1 5 - 6 605 103
GenBank mRNA AF319616.1 - 5 - 6 627 103
AF328908.1 - 5 - 6 636 103

View Full Product Details