Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-56
Entrez Gene ID
188073
Gene Name
Putative UDP-glucuronosyltransferase ugt-56
Gene Aliases
CELE_T04H1.8
Chromosome Location
Chr.V: 12258982 - 12261215 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073673.6 NP_506074.2 1 - 2 167 78

View Full Product Details