Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpa-13
Entrez Gene ID
179746
Gene Name
protein-coding
Gene Aliases
CELE_F18E2.5
Chromosome Location
Chr.V: 12752603 - 12754939 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269521.2 NP_001256450.1 7 - 8 875 96
NM_001269522.3 NP_001256451.1 8 - 9 1083 96
GenBank mRNA AY008135.1 - 7 - 8 875 96

View Full Product Details