Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ril-1
Entrez Gene ID
179958
Gene Name
RNAi-Induced Longevity
Gene Aliases
CELE_C53A5.1
Chromosome Location
Chr.V: 14523330 - 14524727 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074197.7 NP_506598.1 2 - 3 254 70

View Full Product Details