Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
obr-2
Entrez Gene ID
180003
Gene Name
protein-coding
Gene Aliases
CELE_F14H8.1
Chromosome Location
Chr.V: 15011870 - 15014264 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129445.5 NP_001122917.1 1 - 2 58 65
NM_074294.5 NP_506695.2 1 - 2 130 65

View Full Product Details