Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-27
Entrez Gene ID
187956
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_T01G5.3
Chromosome Location
Chr.V: 15126212 - 15127513 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074329.1 NP_506730.1 4 - 5 447 66

View Full Product Details