Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sri-25
Entrez Gene ID
183386
Gene Name
Serpentine Receptor, class I
Gene Aliases
CELE_C41G6.10
Chromosome Location
Chr.V: 15217005 - 15219320 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074359.2 NP_506760.1 4 - 5 932 84

View Full Product Details