Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
twk-34
Entrez Gene ID
180054
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_K06B4.12
Chromosome Location
Chr.V: 15704960 - 15709592 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074505.5 NP_506906.3 6 - 7 1104 146

View Full Product Details